Merck
  • 2,5-Dimethyl-celecoxib inhibits cell cycle progression and induces apoptosis in human leukemia cells.
2,5-Dimethyl-celecoxib inhibits cell cycle progression and induces apoptosis in human leukemia cells.
The Journal of pharmacology and experimental therapeutics (2015-09-04)
Cyril Sobolewski, Jiyun Rhim, Noémie Legrand, Florian Muller, Claudia Cerella, Fabienne Mack, Sébastien Chateauvieux, Jeoung-Gyun Kim, Ah-Young Yoon, Kyu-Won Kim, Mario Dicato, Marc Diederich
ABSTRACT
Cyclooxygenase-2 (COX-2) is an essential regulator of cancer promotion and progression. Extensive efforts to target this enzyme have been developed to reduce growth of cancer cells for chemopreventive and therapeutic reasons. In this context, cyclooxygenase-2 inhibitors present interesting antitumor effects. However, inhibition of COX-2 by anti-COX-2 compounds such as celecoxib was recently associated with detrimental cardiovascular side effects limiting their clinical use. As many anticancer effects of celecoxib are COX-2 independent, analogs such as 2,5-dimethyl-celecoxib (DMC), which lacks COX-2-inhibitory activity, represent a promising alternative strategy. In this study, we investigated the effect of this molecule on growth of hematologic cancer cell lines (U937, Jurkat, Hel, Raji, and K562). We found that this molecule is able to reduce the growth and induces apoptosis more efficiently than celecoxib in all the leukemic cell lines tested. Cell death was associated with downregulation of Mcl-1 protein expression. We also found that DMC induces endoplasmic reticulum stress, which is associated with a decreased of GRP78 protein expression and an alteration of cell cycle progression at the G1/S transition in U937 cells. Accordingly, typical downregulation of c-Myc and cyclin D1 and an upregulation of p27 were observed. Interestingly, for shorter time points, an alteration of mitotic progression, associated with the downregulation of survivin protein expression was observed. Altogether, our data provide new evidence about the mode of action of this compound on hematologic malignancies.
Sigma-Aldrich
Ethylene, ≥99.5%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SAFC
L-Glutamine
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent