Merck
  • Amino acid analysis using chromatography-mass spectrometry: An inter platform comparison study.
Amino acid analysis using chromatography-mass spectrometry: An inter platform comparison study.
Journal of pharmaceutical and biomedical analysis (2015-06-27)
P Krumpochova, B Bruyneel, D Molenaar, A Koukou, M Wuhrer, W M A Niessen, M Giera
ABSTRACT
The analysis of amino acids has become a central task in many aspects. While amino acid analysis has traditionally mainly been carried out using either gas chromatography (GC) in combination with flame ionization detection or liquid chromatography (LC) with either post-column derivatization using ninhydrin or pre-column derivatization using o-phthalaldehyde, many of today's analysis platforms are based on chromatography in combination with mass spectrometry (MS). While derivatization is mandatory for the GC-based analysis of amino acids, several LC platforms have emerged, particularly in the dawn of targeted metabolite profiling using hydrophilic interaction liquid chromatography (HILIC) coupled to MS, allowing the analysis of underivatized amino acids. Among the numerous analytical platforms available for amino acid analysis today, we here compare three prominent approaches, being GC-MS and LC-MS after amino acid derivatization using chloroformate and HILIC-MS of underivatized amino acids. We compare and discuss practical issues as well as performance characteristics, e.g., the use of (13)C-labeled internal standards, of the different platforms and present data on their practical implementation in our laboratory. Finally, we compare the real-life applicability of all three platforms for a complex biological sample. While all three platforms are very-well suited for the analysis of complex biological samples they all show advantages and disadvantages for some analytes as discussed in detail in this manuscript.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
SAFC
L-Threonine
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
DL-Histidine, ≥99% (TLC)
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
DL-Methionine, ≥99%
SAFC
L-Isoleucine
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
DL-Norvaline
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG