Merck
  • Multiplexed tracking of combinatorial genomic mutations in engineered cell populations.
Multiplexed tracking of combinatorial genomic mutations in engineered cell populations.
Nature biotechnology (2015-03-24)
Ramsey I Zeitoun, Andrew D Garst, George D Degen, Gur Pines, Thomas J Mansell, Tirzah Y Glebes, Nanette R Boyle, Ryan T Gill
ABSTRACT
Multiplexed genome engineering approaches can be used to generate targeted genetic diversity in cell populations on laboratory timescales, but methods to track mutations and link them to phenotypes have been lacking. We present an approach for tracking combinatorial engineered libraries (TRACE) through the simultaneous mapping of millions of combinatorially engineered genomes at single-cell resolution. Distal genomic sites are assembled into individual DNA constructs that are compatible with next-generation sequencing strategies. We used TRACE to map growth selection dynamics for Escherichia coli combinatorial libraries created by recursive multiplex recombineering at a depth 10(4)-fold greater than before. TRACE was used to identify genotype-to-phenotype correlations and to map the evolutionary trajectory of two individual combinatorial mutants in E. coli. Combinatorial mutations in the human ES2 ovarian carcinoma cell line were also assessed with TRACE. TRACE completes the combinatorial engineering cycle and enables more sophisticated approaches to genome engineering in both bacteria and eukaryotic cells than are currently possible.
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
2-Methyl-1-propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Methyl-1-propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Isobutyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Isobutyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture