Merck
  • Small-Anion Selective Transmembrane "Holes" Induced by an Antimicrobial Peptide Too Short to Span Membranes.
Small-Anion Selective Transmembrane "Holes" Induced by an Antimicrobial Peptide Too Short to Span Membranes.
The journal of physical chemistry. B (2015-07-01)
Kan Hu, Yunjiang Jiang, Yuntao Xie, Hui Liu, Rui Liu, Zhi Zhao, Ren Lai, Lihua Yang
ABSTRACT
Whereas many membrane-destabilization modes have been suggested for membrane-spanning antimicrobial peptides (AMPs), few are available for those too short to span membrane thickness. Here we show that ORB-1, a 15-residue disulfide-bridged AMP that is only ∼20 Å long even when fully stretched like a hairpin, may act by inducing small anion-selective transmembrane "holes" of negative mean curvature. In model membranes of Gram-negative bacteria, ORB-1 induces chloride transmembrane transport and formation of transmembrane channels of negative mean curvature, whereas the inactive analogue, ORB-N, does not, suggesting a correlation between antibacterial activity and ability to induce transmembrane channels. Given that ORB-N is the C-terminus amidated form of ORB-1, our results further suggest that formation of membrane-spanning dimers may be required to initiate the observed channel induction. Moreover, ORB-1 renders model bacterial membranes permeable to anions with effective hydration diameters of <1 nm (e.g., Cl(-) and NO3(-)), but not cations of similar sizes (e.g., H3O(+)), indicative of anion-selective transmembrane channels with an effective inner diameter of ≤1 nm. In addition, negative-intrinsic-curvature (NIC) lipids such as phosphoethanolamine (PE) may facilitate the membrane-destabilization process of ORB-1. Our findings may expand current understandings on how AMPs destabilize membranes and facilitate the pharmaceutical development of ORB-1.
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥99.0% (10 mg phospholipid per ml CHCl3, TLC)
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
N,N′-Dimethyl-9,9′-biacridinium dinitrate, used as chemiluminescent reagent
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, ≥97.0% (TLC)