Merck
  • Unraveling the functions of type II-prohibitins in Arabidopsis mitochondria.
Unraveling the functions of type II-prohibitins in Arabidopsis mitochondria.
Plant molecular biology (2015-04-22)
Janusz Piechota, Monika Bereza, Aleksandra Sokołowska, Kondrad Suszyński, Karolina Lech, Hanna Jańska
ABSTRACT
In yeast and mammals, prohibitins (PHBs) are considered as structural proteins that form a scaffold-like structure for interacting with a set of proteins involved in various processes occurring in the mitochondria. The role of PHB in plant mitochondria is poorly understood. In the study, the model organism Arabidopsis thaliana was used to identify the possible roles of type-II PHBs (homologs of yeast Phb2p) in plant mitochondria. The obtained results suggest that the plant PHB complex participates in the assembly of multisubunit complexes; namely, respiratory complex I and enzymatic complexes carrying lipoic acid as a cofactor (pyruvate dehydrogenase, 2-oxoglutarate dehydrogenase and glycine decarboxylase). PHBs physically interact with subunits of these complexes. Knockout of two Arabidopsis type-II prohibitins (AtPHB2 and AtPHB6) results in a decreased abundance of these complexes along with a reduction in mitochondrial acyl carrier proteins. Also, the absence of AtPHB2 and AtPHB6 influences the expression of the mitochondrial genome and leads to the activation of alternative respiratory pathways, namely alternative oxidase and external NADH-dependent alternative dehydrogenases.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%