Merck
  • Effect of propylene glycol on adipose tissue mobilization in postpartum over-conditioned Holstein cows.
Effect of propylene glycol on adipose tissue mobilization in postpartum over-conditioned Holstein cows.
Journal of dairy science (2015-10-12)
V Bjerre-Harpøth, A C Storm, M Eslamizad, B Kuhla, M Larsen
ABSTRACT
Our objective was to investigate the quantitative and qualitative effects of propylene glycol (PG) allocation on postpartum adipose tissue mobilization in over-conditioned Holstein cows. Nine ruminally cannulated and arterially catheterized cows were, at parturition, randomly assigned to a ruminal pulse dose of either 500g of tap water (n=4) or 500g of PG (n=5) once a day. The PG was given with the morning feeding for 4 wk postpartum (treatment period), followed by a 4-wk follow-up period. All cows were fed the same prepartum and postpartum diets. At -16 (±3), 4 (±0), 15 (±1) and 29 (±2) days in milk (DIM), body composition was determined using the deuterium oxide dilution technique, liver and subcutaneous adipose tissue biopsies were collected, and mammary gland nutrient uptake was measured. Weekly blood samples were obtained during the experiment and daily blood samples were taken from -7 to 7 DIM. Postpartum feed intake and milk yield was not affected by PG allocation. The body content of lipid was not affected by treatment, but tended to decrease from 4 to 29 DIM with both treatments. Except for the first week postpartum, no difference in plasma nonesterified fatty acids concentration was noted between treatments in the treatment period. Yet, PG allocation resulted in decreased plasma concentrations of β-hydroxybutyrate (BHB) and increased plasma concentrations of glucose. In the follow-up period, plasma concentrations of nonesterified fatty acids, glucose, and BHB did not differ between treatments. Additionally, the change in abundance of proteins in adipose tissue biopsies from prepartum to 4 DIM was not affected by treatment. In conclusion, the different variables to assess body fat mobilization were concurrent and showed that a 4-wk postpartum PG allocation had limited effect on adipose tissue mobilization. The main effect was an enhanced glucogenic status with PG. No carry-over effect of PG allocation was recorded for production or plasma metabolites, and, hence, a new period of metabolic adaption to lactation seemed to occur with PG treatment after ceasing PG allocation. Thus, PG seemed to induce a 2-step adaption to lactation, reducing the immediate postpartum nadir and peak of plasma concentration of glucose and BHB, respectively; which is beneficial for postpartum cows at high risk of lipid-related metabolic diseases.
Sigma-Aldrich
3,3′-Diindolylmethane, ≥98% (HPLC)
Sigma-Aldrich
Glucagon, ≥95% (HPLC), powder, synthetic
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Glucagon, synthetic, powder, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
Urea, meets USP testing specifications