Merck
  • Differences in the glucuronidation of bisphenols F and S between two homologous human UGT enzymes, 1A9 and 1A10.
Differences in the glucuronidation of bisphenols F and S between two homologous human UGT enzymes, 1A9 and 1A10.
Xenobiotica; the fate of foreign compounds in biological systems (2014-12-31)
Darja Gramec Skledar, Johanna Troberg, Jason Lavdas, Lucija Peterlin Mašič, Moshe Finel
ABSTRACT
1. Bisphenol S (BPS) and bisphenol F (BPF) are bisphenol A (BPA) analogues commonly used in the manufacturing of industrial and consumer products. 2. Bisphenols are often detoxified through conjugation with glucuronic acid or sulfate. In this work, we have examined the glucuronidation of BPS and BPF by recombinant human UDP-glucuronosyltransferase (UGT) enzymes. In addition, we have reexamined BPA glucuronidation, using extra-hepatic UGTs that were not tested previously. 3. The results revealed that UGT1A9, primarily a hepatic enzyme, is mainly responsible for BPS glucuronidation, whereas UGT1A10, an intestine enzyme that is highly homologous to UGT1A9 at the protein level, is by far the most active UGT in BPF glucuronidation. In contrast to the latter two UGTs that display significant specificity in the glucuronidation of BPS and BPF, UGT2A1 that is mainly expressed in the airways, exhibited high activity toward all the tested bisphenols, BPS, BPF and BPA. UGT1A10 exhibited somewhat higher BPA glucuronidation activity than UGT1A9, but it was lower than UGT2A1 and UGT2B15. 4. The new findings demonstrate interesting differences in the glucuronidation patterns of bisphenols and provide new insights into the role of extra-hepatic tissues in their detoxification.
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, Molecular Biology, ≥99.0% (T)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Magnesium chloride hexahydrate, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Magnesium chloride hexahydrate, 99.995% trace metals basis
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride hexahydrate, meets USP testing specifications
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%