Merck
  • The augmentation of O-GlcNAcylation reduces glyoxal-induced cell injury by attenuating oxidative stress in human retinal microvascular endothelial cells.
The augmentation of O-GlcNAcylation reduces glyoxal-induced cell injury by attenuating oxidative stress in human retinal microvascular endothelial cells.
International journal of molecular medicine (2015-08-28)
Guo Dong Liu, Chong Xu, Le Feng, Fang Wang
ABSTRACT
It has recently been reported that O-linked β-N-acetyl glucosamine (O-GlcNAc) modification (a simple intracellular serine (Ser)/threonine (Thr)-linked monosaccharide) in human retinal microvascular endothelial cells (HRECs) is related to diabetic retinopathy (DR). During O-GlcNAcylation, O-GlcNAc is added to Ser and Thr residues. As the generation of reactive oxygen species (ROS) is one of the characteristics of advanced glycation end product (AGE) injury, and the most important key pathogenic factor of DR, in the present study, we aimed to investigate the association between O-GlcNAcylation and ROS generation in order to ascertain whether O-GlcNAcylation mitigates cellular injury through the generation of ROS. For this purpose, HRECs were divided into 4 groups as follows: HRECs treated with normal glucose (5 mM), HRECs treated with glyoxal (500 µM), glyoxal-treated HRECs also treated with 200 µM PUGNAc, and glyoxal-treated HRECs infected with O-GlcNAc transferase (OGT) siRNA. We detected increased O-GlcNAc levels and increased ROS production in the glyoxal-treated HRECs. The cellular redox status was determined by cellular ROS staining and by measuring the expression levels of the antioxidant genes, superoxide dismutase (SOD) and glutathione peroxidase (GPX). While the augmentation of O-GlcNAcylation following treatment with PUGNAc significantly attenuated the production of ROS (p<0.01) and increased the expression levels of SOD and GPX, the reduction of O-GlcNAcylation following infection with OGT siRNA, exacerbated the production of ROS (p<0.01) and decreased the expression of antioxidant genes. The effects of O-GlcNAcylation on the viability of HRECs were significant (p<0.01), particularly in the hydrogen peroxide (H2O2)-treated HRECs. Treatment with PUGNAc reduced glyoxal-induced cell apoptosis and transfection with OGT siRNA increased HREC apoptosis; these results were confirmed by flow cytometry and by the assessment of mitochondrial membrane potential. The augmentation of O-GlcNAcylation exerted cytoprotective effects on the HRECs by reducing the generation of ROS, increasing the expression of antioxidant genes, preventing the dissipation of mitochondrial membrane potential and preventing HREC apoptosis. Therefore, it can be concluded that O-GlcNAcylation plays a role in the early developmental process of DR.
Product Description
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Glyoxal solution, 40 wt. % in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Glyoxal solution, BioReagent, ~40% in H2O (~8.8 M), Molecular Biology
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ogt
Sigma-Aldrich
MISSION® esiRNA, targeting human OGT
Sigma-Aldrich
JC-1, powder or solid (Crystals)