Merck
  • Structure characterization of unexpected covalent O-sulfonation and ion-pairing on an extremely hydrophilic peptide with CE-MS and FT-ICR-MS.
Structure characterization of unexpected covalent O-sulfonation and ion-pairing on an extremely hydrophilic peptide with CE-MS and FT-ICR-MS.
Analytical and bioanalytical chemistry (2015-07-01)
Martin Pattky, Simone Nicolardi, Beatrix Santiago-Schübel, Daniel Sydes, Yuri E M van der Burgt, Antonia N Klein, Nan Jiang, Jeannine Mohrlüder, Karen Hänel, Janine Kutzsche, S A Funke, D Willbold, S Willbold, C Huhn
ABSTRACT
In this study, we characterized unexpected side-products in a commercially synthesized peptide with the sequence RPRTRLHTHRNR. This so-called peptide D3 was selected by mirror phage display against low molecular weight amyloid-β-peptide (Aβ) associated with Alzheimer's disease. Capillary electrophoresis (CE) was the method of choice for structure analysis because the extreme hydrophilicity of the peptide did not allow reversed-phase liquid chromatography (RPLC) and hydrophilic interaction stationary phases (HILIC). CE-MS analysis, applying a strongly acidic background electrolyte and different statically adsorbed capillary coatings, provided fast and efficient analysis and revealed that D3 unexpectedly showed strong ion-pairing with sulfuric acid. Moreover, covalent O-sulfonation at one or two threonine residues was identified as a result of a side reaction during peptide synthesis, and deamidation was found at either the asparagine residue or at the C-terminus. In total, more than 10 different species with different m/z values were observed. Tandem-MS analysis with collision induced dissociation (CID) using a CE-quadrupole-time-of-flight (QTOF) setup predominantly resulted in sulfate losses and did not yield any further characteristic fragment ions at high collision energies. Therefore, direct infusion Fourier transform ion cyclotron resonance (FT-ICR) MS was employed to identify the covalent modification and discriminate O-sulfonation from possible O-phosphorylation by using an accurate mass analysis. Electron transfer dissociation (ETD) was used for the identification of the threonine O-sulfation sites. In this work, it is shown that the combination of CE-MS and FT-ICR-MS with ETD fragmentation was essential for the full characterization of this extremely basic peptide with labile modifications.
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
N-Acetyl-D-lactosamine, ≥98% (TLC)
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
SAFC
Iodoacetamide
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG