Merck
  • Calea zacatechichi dichloromethane extract exhibits antidiarrheal and antinociceptive effects in mouse models mimicking irritable bowel syndrome.
Calea zacatechichi dichloromethane extract exhibits antidiarrheal and antinociceptive effects in mouse models mimicking irritable bowel syndrome.
Naunyn-Schmiedeberg's archives of pharmacology (2015-06-13)
M Sałaga, A Kowalczuk, M Zielinska, A Błażewicz, J Fichna
ABSTRACT
Calea zacatechichi Schltdl. (Asteraceae alt. Compositae) is a Mexican plant commonly used in folk medicine to treat respiratory and gastrointestinal (GI) disorders. The objective of this study is to characterize the effect of C. zacatechichi extracts in mouse models mimicking the symptoms of irritable bowel syndrome (IBS). Powdered C. zacatechichi herb (leaves, stems, and flowers) was extracted with methanol. Methanolic extract was filtered and evaporated giving methanolic fraction. The residue was extracted with dichloromethane (DCM). Methanolic and DCM (200 mg/kg, per os) extracts were screened for their effect on GI motility in several in vitro tests, and the antidiarrheal and antinociceptive effects were assessed using mouse models. The influence of the DCM extract on motoric parameters and exploratory behaviors was also assessed. Finally, the composition of C. zacatechichi DCM extract was qualitatively analyzed using liquid chromatography-mass spectrometry (LC-MS) method. C. zacatechichi DCM extract significantly inhibited the contractility of mouse colon in vitro (IC50 = 17 ± 2 μg/ml). Administration of the DCM extract in vivo (200 mg/kg, per os) significantly prolonged the time of whole GI transit (46 ± 1 vs. 117 ± 27 min for control and DCM-treated animals, respectively; P = 0.0023), inhibited hypermotility, and reduced pain in mouse models mimicking functional GI disorders. Our findings suggest that constituents of the C. zacatechichi DCM extract exhibit antidiarrheal and analgesic activity. The extract may thus become an attractive material for isolation of compounds that may be used as a supplementary treatment for pain and diarrhea associated with IBS in the future.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis