Merck
  • Antioxidant-mediated reversal of oxidative damage in mouse modeling of complex I inhibition.
Antioxidant-mediated reversal of oxidative damage in mouse modeling of complex I inhibition.
Drug development research (2015-04-08)
Kodeeswaran Parameshwaran, Michael H Irwin, Kosta Steliou, Vishnu Suppiramaniam, Carl A Pinkert
ABSTRACT
Mitochondrial dysfunction is a key component of various aging-related pathologies of the brain that result in dementia. As such, it provides an important avenue in development of therapeutic interventions for a host of neurological disorders. A requirement for functional mitochondrial respiratory chain complex I (CI), to accomplish the normal physiological processes regulating memory, seems intuitive. In the present study, a synthetic lipoylcarnitine antioxidant (PMX-500FI; 100 mg/kg/day po) was administered to female ICR mice (3-4-month old) that were subsequently treated with the mitochondrial CI inhibitor, rotenone (400 mg/kg/day). After 1 week, rotenone-induced impairment of neuronal function was evaluated in the hippocampus, a brain region that is involved in regulating memory formation. Electrophysiological recordings in live brain slices showed that long-term potentiation (LTP) was reduced by rotenone exposure (P < 0.05) while pretreatment with PMX-500FI maintained LTP similar to control levels (P > 0.05). Potentiation during theta burst stimulation (TBS) was similar among treatment groups (P > 0.05); however, neurotransmitter release, which increased in control mice after TBS, was lower in rotenone treated mice (P < 0.05), and was accompanied by reduced basal synaptic transmission (P < 0.05), increased proapoptotic signaling and decreased extracellular signal-regulated kinase1/2 (ERK1/2) phosphorylation (P < 0.05). For each of these determinations, pretreatment with PMX-500FI alleviated the harmful effects of rotenone. These results illustrate that treatment with antioxidant PMX-500FI is protective against rotenone-induced impairment of neuronal bioenergetics in the mouse hippocampus, in regard to both excitatory synaptic physiology and proapoptotic signaling. The protective effect of PMX-500FI against rotenone-induced disruption of cellular bioenergetics may have important therapeutic implications for treating aging-related dementia and other diseases related to mitochondrial dysfunction and/or oxidative damage.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent