Merck
  • Characteristics of morphology, electrophysiology, and central projections of two sensilla styloconica in Helicoverpa assulta larvae.
Characteristics of morphology, electrophysiology, and central projections of two sensilla styloconica in Helicoverpa assulta larvae.
Neuroreport (2015-07-15)
Qing-Bo Tang, Zhen-Zhen Hong, Huan Cao, Feng-Ming Yan, Xin-Cheng Zhao
ABSTRACT
The medial and lateral styloconic sensilla, constituting the main taste organs of lepidopterous caterpillars, were investigated in the oligophagous species, Helicoverpa assulta (Guenée) (Lepidoptera: Noctuidae). In this paper, the two sensilla were morphologically and physiologically characterized by scanning electron microscopy and tip recordings, respectively. The central projections of their respective sensory neurons were mapped by anterograde staining experiments combined with confocal laser scanning microscopy. The results showed that the two sensilla are in general morphologically similar. However, the size of the peg on the medial sensillum is significantly greater than that of the lateral. Tobacco leaf saps, sinigrin, and nicotine elicited strong responses from neurons housed by the medial sensillum, whereas sucrose activated primarily the lateral sensillum. All stained neurons in either sensillum showed a projection pattern involving axons entering the subesophageal ganglion through the ipsilateral maxillary and passing further on through the ipsilateral circumesophageal connective to the tritocerebrum of the brain. In the subesophageal ganglion, the axons targeted two areas: the ventrolateral section and the region near the neuromere midline. One distinction between the staining patterns originating from the two sensilla, however, is that axons arising from the medial sensillum, and not the lateral, give off some additional neural branches in the subesophageal ganglion including a few arborizations surrounding a tract, plus a long process extending posteriorly along the midline. Differences in the central projections derived from the two sensilla styloconica have not been reported previously.
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%