Merck
  • Regular postexercise cooling enhances mitochondrial biogenesis through AMPK and p38 MAPK in human skeletal muscle.
Regular postexercise cooling enhances mitochondrial biogenesis through AMPK and p38 MAPK in human skeletal muscle.
American journal of physiology. Regulatory, integrative and comparative physiology (2015-06-05)
Mohammed Ihsan, James F Markworth, Greig Watson, Hui Cheng Choo, Andrew Govus, Toan Pham, Anthony Hickey, David Cameron-Smith, Chris R Abbiss
ABSTRACT
This study investigated the effect of regular postexercise cold water immersion (CWI) on muscle aerobic adaptations to endurance training. Eight males performed 3 sessions/wk of endurance training for 4 wk. Following each session, subjects immersed one leg in a cold water bath (10°C; COLD) for 15 min, while the contralateral leg served as a control (CON). Muscle biopsies were obtained from vastus lateralis of both CON and COLD legs prior to training and 48 h following the last training session. Samples were analyzed for signaling kinases: p38 MAPK and AMPK, peroxisome proliferator-activated receptor gamma coactivator-1α (PGC-1α), enzyme activities indicative of mitochondrial biogenesis, and protein subunits representative of respiratory chain complexes I-V. Following training, subjects' peak oxygen uptake and running velocity were improved by 5.9% and 6.2%, respectively (P < 0.05). Repeated CWI resulted in higher total AMPK, phosphorylated AMPK, phosphorylated acetyl-CoA carboxylase, β-3-hydroxyacyl-CoA-dehydrogenase and the protein subunits representative of complex I and III (P < 0.05). Moreover, large effect sizes (Cohen's d > 0.8) were noted with changes in protein content of p38 (d = 1.02, P = 0.064), PGC-1α (d = 0.99, P = 0.079), and peroxisome proliferator-activated receptor α (d = 0.93, P = 0.10) in COLD compared with CON. No differences between conditions were observed in the representative protein subunits of respiratory complexes II, IV, and V and in the activities of several mitochondrial enzymes (P > 0.05). These findings indicate that regular CWI enhances p38, AMPK, and possibly mitochondrial biogenesis.
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-PGC-1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Citrate Synthase from porcine heart, liquid, ≥100 units/mg protein
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.85%