Merck
  • Restoration of Wnt/β-catenin signaling attenuates alcoholic liver disease progression in a rat model.
Restoration of Wnt/β-catenin signaling attenuates alcoholic liver disease progression in a rat model.
Journal of hepatology (2015-03-01)
Chiung-Kuei Huang, Tunan Yu, Suzanne M de la Monte, Jack R Wands, Zoltan Derdak, Miran Kim
ABSTRACT
Alcoholic liver disease (ALD) is characterized by the development of fatty liver, alcoholic hepatitis, fibrosis and cirrhosis. However, the underlying mechanism(s) associated with progression remains elusive. Pro-inflammatory cytokines have been implicated in ALD progression due to pro-apoptotic effects on hepatocytes. Wnt/β-catenin signaling recently has been shown to promote inflammation and apoptosis, suggesting that activation of this signaling pathway may modulate ALD progression. The current study was designed to test whether pharmacological activation of Wnt/β-catenin signaling altered ALD development and progression in a rat model. Adult male Long Evans rats were fed with isocaloric liquid diets containing 0% or 37% ethanol for 8 weeks, and also treated with Wnt agonist during the last 3 weeks of the feeding regimen. Liver and blood samples were subjected to histology, TUNEL assay, immunoblot analysis, real-time quantitative PCR, and alanine transaminase (ALT) assay. Wnt/β-catenin signaling was negatively correlated with Foxo3A expression and reduced steatosis, cellular injury and apoptosis in ALD rats. Mutation experiments demonstrated that Foxo3A was critical for modulating these effects. Activation of Wnt/β-catenin signaling suppressed Foxo3A-induced apoptosis through upregulation of serum/glucocorticoid regulated kinase 1 (SGK1). Moreover, pharmacological restoration of Wnt/β-catenin signaling reduced ALD progression in vivo. Wnt/β-catenin signaling plays a protective role in ALD progression via antagonizing Foxo3A-induced apoptosis, and activation of the Wnt/β-catenin signaling cascade attenuates ALD progression.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
SAFC
L-Glutamine
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology