Merck
  • Application of a compact magnetic resonance imaging system for toxicologic pathology: evaluation of lithium-pilocarpine-induced rat brain lesions.
Application of a compact magnetic resonance imaging system for toxicologic pathology: evaluation of lithium-pilocarpine-induced rat brain lesions.
Journal of toxicologic pathology (2015-11-06)
Yoshikazu Taketa, Motohiro Shiotani, Yoshiharu Tsuru, Sadaharu Kotani, Yoshihide Osada, Tatsuto Fukushima, Akira Inomata, Satoru Hosokawa
ABSTRACT
Magnetic resonance imaging (MRI) is a useful noninvasive tool used to detect lesions in clinical and veterinary medicine. The present study evaluated the suitability of a new easy-to-use compact MRI platform (M2 permanent magnet system, Aspect Imaging, Shoham, Israel) for assisting with preclinical toxicologic pathology examination of lesions in the rat brain. In order to induce brain lesions, male Sprague-Dawley rats were treated once with lithium chloride (127 mg/kg, intraperitoneal [i.p.]) followed by pilocarpine (30 mg/kg, i.p.). One week after dosing, the perfused, fixed brains were collected, analyzed by the MRI system and examined histopathologically. MRI of the brain of treated rats revealed areas of high T1 and middle to low T2 signals, when compared with the controls, in the piriform cortex, lateral thalamic nucleus, posterior paraventricular thalamic nucleus and posterior hypothalamic nucleus of the cerebrum. The altered MRI signal areas were consistent with well-circumscribed foci of neuronal cell degeneration/necrosis accompanied by glial cell proliferation. The present data demonstrated that quick analysis of fixed organs by the MRI system can detect the presence and location of toxicologic lesions and provide useful temporal information for selection of appropriate sections for histopathologic examination before routine slide preparation, especially in complex and functionally heterogeneous organs such as the brain.
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%