Merck
  • Cholesterol anchored arabinogalactan for asialoglycoprotein receptor targeting: synthesis, characterization, and proof of concept of hepatospecific delivery.
Cholesterol anchored arabinogalactan for asialoglycoprotein receptor targeting: synthesis, characterization, and proof of concept of hepatospecific delivery.
Carbohydrate research (2015-04-05)
Pankaj Omprakash Pathak, Mangal Shailesh Nagarsenker, Chandrashekhar Rishikant Barhate, Sameer Govind Padhye, Vivek Vijay Dhawan, Dibyendu Bhattacharyya, C L Viswanathan, Frank Steiniger, Alfred Fahr
ABSTRACT
Asialoglycoprotein receptors (ASGPR) are hepatocyte bound receptors, which exhibit receptor mediated endocytosis (RME) for galactose specific moieties. Arabinogalactan (AG), a liver specific high galactose containing branched polysaccharide was hydrophobized using cholesterol (CHOL) as a lipid anchor via a two step reaction process to yield the novel polysaccharide lipid conjugated ligand (CHOL-AL-AG). CHOL-AL-AG was characterized by Fourier transform infra red (FTIR) spectroscopy, (1)H and (13)C nuclear magnetic spectroscopy (NMR), size exclusion chromatography (SEC) and differential scanning calorimetry (DSC). Conventional liposomes (CL) and surface modified liposomes (SML) containing CHOL-AL-AG were prepared using reverse phase evaporation technique. Effect of CHOL-AL-AG concentration on particle size and zeta potential of SML was evaluated. Surface morphology of CL and SML was studied using cryo-transmission electron microscopy (cryo-TEM). In vitro binding affinity of SML and CL was evaluated using Ricinus communis agglutinin (RCA) assay. Cellular uptake of SML and CL was determined on ASGPR expressing HepG2 cell lines by confocal laser scanning microscopy technique (CLSM). FTIR spectra revealed bands at 1736 cm(-1) and 1664 cm(-1) corresponding to ester and carbamate functional groups, respectively. Signals at δ 0.5-2.5 corresponding to the cholestene ring and δ 3-5.5 corresponding to the carbohydrate backbone were observed in (1)H NMR spectrum of the product. CHOL-AL-AG possessed a mean average molecular weight of 27 KDa as determined by size exclusion chromatography. An endothermic peak at 207 °C was observed in the DSC thermogram of CHOL-AL-AG, which was not observed in thermograms of reactants and intermediate product. Synthesized CHOL-AL-AG was successfully incorporated in liposomes to yield SML. Both CL and SML possessed a mean particle size of ∼ 200 nm with polydispersity index of ∼ 0.25. The zeta potential of CLs was observed to be -17 mV whereas zeta potential of SMLs varied from -18 to -22 mV. RCA assay revealed enhanced binding of SML compared to CL confirming presence of galactose on surface of SML. CLSM studies demonstrated enhanced cellular uptake of SMLs compared to CL by HepG2 cells post 3 h administration indicating enhanced uptake by the ASGPR. Thus surface modified liposomes specific to target heptocytes demonstrate a promising approach for targeted drug delivery in liver cancer therapeutics.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
CDI, ≥97.0% (T)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
CDI, reagent grade
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Cholesteryl chloroformate, 95%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%