Merck
  • Development and characterization of chitosan/hyaluronan film for transdermal delivery of thiocolchicoside.
Development and characterization of chitosan/hyaluronan film for transdermal delivery of thiocolchicoside.
Carbohydrate polymers (2015-06-17)
Federica Bigucci, Angela Abruzzo, Bruno Saladini, Maria Caterina Gallucci, Teresa Cerchiara, Barbara Luppi
ABSTRACT
The objective of this study was the development of chitosan/hyaluronan transdermal films to improve bioavailability of thiocolchicoside. This approach offers the possibility to elude the first-pass metabolism and at the same time it is able to provide a predictable and extended duration of activity. Films were prepared by casting and drying of aqueous solutions containing different weight ratios of chitosan and hyaluronan and characterized for their physico-chemical and functional properties. In accordance with polymeric composition of films and, therefore, with the amount of the net charge after the complexation, films containing the same weight ratio of chitosan and hyaluronan showed lower water uptake ability with respect to films containing only one polymeric species or an excess of chitosan or hyaluronan. Moreover, the lower the hydration of the polymeric network, the lower is the drug diffusion through the films and its permeation through the skin. This study clearly confirmed that the selection of a suitable polymeric weight ratio and appropriate preparative conditions allows the modulation of film functional properties, suggesting that these formulations could be used as a novel technological platform for transdermal drug delivery.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis