Merck
  • Drug Redeployment to Kill Leukemia and Lymphoma Cells by Disrupting SCD1-Mediated Synthesis of Monounsaturated Fatty Acids.
Drug Redeployment to Kill Leukemia and Lymphoma Cells by Disrupting SCD1-Mediated Synthesis of Monounsaturated Fatty Acids.
Cancer research (2015-05-07)
Andrew D Southam, Farhat L Khanim, Rachel E Hayden, Julia K Constantinou, Katarzyna M Koczula, Robert H Michell, Mark R Viant, Mark T Drayson, Chris M Bunce
ABSTRACT
The redeployed drug combination of bezafibrate and medroxyprogesterone acetate (designated BaP) has potent in vivo anticancer activity in acute myelogenous leukemia (AML) and endemic Burkitt lymphoma (eBL) patients; however, its mechanism-of-action is unclear. Given that elevated fatty acid biosynthesis is a hallmark of many cancers and that these drugs can affect lipid metabolism, we hypothesized that BaP exerts anticancer effects by disrupting lipogenesis. We applied mass spectrometry-based lipidomics and gene and protein expression measurements of key lipogenic enzymes [acetyl CoA carboxylase 1 (ACC1), fatty acid synthase (FASN), and stearoyl CoA desaturase 1 (SCD1)] to AML and eBL cell lines treated with BaP. BaP treatment decreased fatty acid and phospholipid biosynthesis from (13)C D-glucose. The proportion of phospholipid species with saturated and monounsaturated acyl chains was also decreased after treatment, whereas those with polyunsaturated chains increased. BaP decreased SCD1 protein levels in each cell line (0.46- to 0.62-fold; P < 0.023) and decreased FASN protein levels across all cell lines (0.87-fold decrease; P = 1.7 × 10(-4)). Changes to ACC1 protein levels were mostly insignificant. Supplementation with the SCD1 enzymatic product, oleate, rescued AML and e-BL cells from BaP cell killing and decreased levels of BaP-induced reactive oxygen species, whereas supplementation with the SCD1 substrate (and FASN product), palmitate, did not rescue cells. In conclusion, these data suggest that the critical anticancer actions of BaP are decreases in SCD1 levels and monounsaturated fatty acid synthesis. To our knowledge, this is the first time that clinically available antileukemic and antilymphoma drugs targeting SCD1 have been reported.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Bezafibrate, ≥98%, solid
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %