Merck
  • Dog erythrocyte antigen 1: mode of inheritance and initial characterization.
Dog erythrocyte antigen 1: mode of inheritance and initial characterization.
Veterinary clinical pathology (2015-08-21)
Klaudia Polak, Michelle M Acierno, Karthik Raj, Keijiro Mizukami, Don L Siegel, Urs Giger
ABSTRACT
The dog erythrocyte antigen (DEA) 1 blood group system remains poorly defined. The purpose of the study was to determine the DEA 1 mode of inheritance and to characterize the DEA 1 antigen and alloantibodies. Canine research colony families, clinic canine patients, and DEA 1.2+ blood bank dogs were studied. Canine blood was typed by flow cytometry and immunochromatographic strips using anti-DEA 1 monoclonal antibodies. Gel column experiments with polyclonal and immunoblotting with monoclonal anti-DEA 1 antibodies were performed to analyze select samples. Cross-reactivity of human typing reagents against canine RBC and one monoclonal anti-DEA 1 antibody against human RBC panels was assessed. Typing of 12 families comprising 144 dogs indicated an autosomal dominant inheritance with ≥ 4 alleles: DEA 1- (0) and DEA 1+ weak (1+), intermediate (2+), and strong (3+ and 4+). Samples from 6 dogs previously typed as DEA 1.2+ were typed as DEA 1+ or DEA 1- using monoclonal antibodies. Human typing reagents produced varied reactions in tube agglutination experiments against DEA 1+ and DEA 1- RBC. Polypeptide bands were not detected on immunoblots using a monoclonal anti-DEA 1 antibody, therefore the anti-DEA 1 antibody may be specific for conformational epitopes lost during processing. The autosomal dominant inheritance of DEA 1 with ≥ 4 alleles indicates a complex blood group system; the antigenicity of each DEA 1+ type will need to be determined. The biochemical nature of the DEA 1 antigen(s) appears different from human blood group systems tested.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
N,N-Diethylaniline, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets