Merck
  • Osmoregulation by juvenile brown-banded bamboo sharks, Chiloscyllium punctatum, in hypo- and hyper-saline waters.
Osmoregulation by juvenile brown-banded bamboo sharks, Chiloscyllium punctatum, in hypo- and hyper-saline waters.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2015-04-15)
R L Cramp, M J Hansen, C E Franklin
ABSTRACT
While there is a considerable body of work describing osmoregulation by elasmobranchs in brackish and saltwater, far fewer studies have investigated osmoregulation in hypersaline waters. We examined osmo- and ionoregulatory function and plasticity in juvenile brown-banded bamboo sharks, Chiloscyllium punctatum, exposed to three experimental salinities (25, 34 and 40‰) for two weeks. C. punctatum inhabits sheltered coastal areas and bays which can naturally become hypersaline as a consequence of evaporation of water but can also become hyposaline during flood events. We hypothesised that C. punctatum would demonstrate a phenotypically plastic osmoregulatory physiology. Plasma osmolality, urea, Na(+) and Cl(-) levels increased significantly with increasing environmental salinity. Rectal gland and branchial sodium-potassium ATPase (NKA) activities were unaffected by salinity. Using immunohistochemistry and Western Blotting we found evidence for the presence of the key ion-regulatory proteins vacuolar H(+)-ATPase (VHA), pendrin (Cl(-)/HCO₃(-) co-transporter) and the Na(+)-H(+) exchanger isoform 3 (NHE3) in discrete cells within the branchial epithelia. These results indicate that C. punctatum is a partially euryhaline elasmobranch able to maintain osmo- and ionoregulatory function between environmental salinities of 25‰ and 40‰. As suggested for other elasmobranchs, the gills of C. punctatum likely play a limited role in maintaining Na(+) homeostasis over the salinity range studied, but may play an important role in acid-base balance.
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C