Merck
  • MMPIP, an mGluR7-selective negative allosteric modulator, alleviates pain and normalizes affective and cognitive behavior in neuropathic mice.
MMPIP, an mGluR7-selective negative allosteric modulator, alleviates pain and normalizes affective and cognitive behavior in neuropathic mice.
Pain (2015-03-12)
Enza Palazzo, Rosaria Romano, Livio Luongo, Serena Boccella, Danilo De Gregorio, Maria Elvira Giordano, Francesca Rossi, Ida Marabese, Maria Antonietta Scafuro, Vito de Novellis, Sabatino Maione
ABSTRACT
This study investigated the effects of a single administration of 6-(4-methoxyphenyl)-5-methyl-3-pyridinyl-4-isoxazolo[4,5-c]pyridin-4(5H)-one (MMPIP), a negative allosteric modulator (NAM) of metabotropic glutamate receptor 7 (mGluR7), on pain and on affective and cognitive behavior in neuropathic mice. The activity of pyramidal neurons in the prelimbic cortex (PLC), which respond to stimulation of the basolateral amygdala (BLA) with either excitation or inhibition, was also investigated. The spared nerve injury (SNI) of the sciatic nerve induced, 14 days after surgery, thermal hyperalgesia and mechanical allodynia, reduced open-arm choice in the elevated plus-maze, increased time of immobility in the tail suspension, and increased digging and burying in the marble burying test. Cognitive performance was also significantly compromised in the SNI mice. Spared nerve injury induced phenotypic changes on pyramidal neurons of the PLC; excitatory responses increased, whereas inhibitory responses decreased after BLA stimulation. mGluR7 expression, mainly associated with vesicular glutamate transporter, increased in the hippocampus and decreased in the BLA, PLC, and dorsal raphe in SNI mice. MMPIP increased thermal and mechanical thresholds and open-arm choice. It reduced the immobility in the tail suspension test and the number of marbles buried and of digging events in the marble burying test. MMPIP also improved cognitive performance and restored the balance between excitatory and inhibitory responses of PLC neurons in SNI mice. 7-hydroxy-3-(4-iodophenoxy)-4H-chromen-4-one, XAP044, another selective mGluR7 NAM, reproduced the effects of MMPIP on thermal hyperalgesia, mechanical allodynia, tail suspension, and marble burying test. Altogether, these findings show that mGluR7 NAMs reduce pain responses and affective/cognitive impairments in neuropathic pain conditions.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Bromophenol Blue, ACS reagent
Sigma-Aldrich
Bromophenol Blue, titration: suitable
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
MMPIP, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%