Merck
  • Short-term exercise training attenuates acute doxorubicin cardiotoxicity.
Short-term exercise training attenuates acute doxorubicin cardiotoxicity.
Journal of physiology and biochemistry (2015-09-26)
Chia-Ying Lien, Brock T Jensen, David S Hydock, Reid Hayward
ABSTRACT
Doxorubicin (DOX) is a potent and widely used antineoplastic agent. Despite the efficacy of DOX, its clinical use is limited by a dose-dependent cardiotoxicity. Chronic exercise training has been shown to protect against DOX-induced cardiotoxicity. It is less clear whether short-term exercise can attenuate DOX-induced dysfunction. The purposes of this study were to determine if short-term wheel running and treadmill exercise training can attenuate the cardiac dysfunction that accompanies DOX treatment and to investigate possible mechanisms that may be involved with any protective effects of exercise. Male Sprague-Dawley rats engaged in a short-term 5-day voluntary wheel running (WR) or treadmill exercise (TM) regimen. Following the exercise preconditioning period, animals received either 10 or 15 mg/kg of DOX or an equivalent volume of saline (SAL). Five days after DOX/SAL exposure, cardiac function was examined. Western immunoblotting was used to quantify left ventricular sarcoendoplasmic reticulum calcium-ATPase 2a (SERCA2a) protein expression. Exercise preconditioning attenuated in vivo and ex vivo cardiac dysfunction observed with DOX treatment alone. Specifically, short-term treadmill exercise (TM + DOX10, 56 ± 4%; TM + DOX15, 48 ± 5%) and voluntary wheel running (WR + DOX10, 51 ± 5%; WR + DOX15, 45 ± 3%) consistently preserved fractional shortening when compared to sedentary (SED) animals treated with DOX (SED + DOX10, 48 ± 4%; SED + DOX15, 39 ± 6%). Likewise, both exercise protocols preserved left ventricular developed pressure (TM + DOX10, 115 ± 6 mmHg; TM + DOX15, 85 ± 5 mmHg; WR + DOX10, 92 ± 12 mmHg; WR + DOX15, 91 ± 8 mmHg) when compared to SED animals treated with DOX (SED + DOX10, 79 ± 6 mmHg; SED + DOX15, 69 ± 7 mmHg). SERCA2a expression was also preserved in TM + DOX and WR + DOX. These findings suggest that short-term exercise prior to DOX treatment may be a valuable adjuvant therapy to offset acute cardiotoxicities and that maintaining calcium handling in cardiomyocytes may be responsible, in part, for the preservation in cardiac function.
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture