Merck
  • Rapid and Sensitive Detection of Vibrio alginolyticus by Loop-Mediated Isothermal Amplification Combined with a Lateral Flow Dipstick Targeted to the rpoX Gene.
Rapid and Sensitive Detection of Vibrio alginolyticus by Loop-Mediated Isothermal Amplification Combined with a Lateral Flow Dipstick Targeted to the rpoX Gene.
Journal of aquatic animal health (2015-07-30)
Saranya Plaon, Siwaporn Longyant, Paisarn Sithigorngul, Parin Chaivisuthangkura
ABSTRACT
Vibrio alginolyticus is a major bacterial pathogen causing disease in marine animals. The present study aimed to develop a loop-mediated isothermal amplification (LAMP) coupled with a lateral flow dipstick (LFD) for rapid and simple visual detection of V. alginolyticus-specific amplicons. The biotin-labeled LAMP amplicons from the targeted portion of a gene encoding rpoS-like sigma factor (rpoX) were generated at 60°C for 1 h and then hybridized with a fluorescein isothiocyanate-labeled probe for 5 min for visual detection with LFD. In pure cultures, the detection limit of the LAMP-LFD technique for V. alginolyticus was 1.8 × 10(2) CFU/mL while that of PCR was 1.8 × 10(3) CFU/mL. In spiked whiteleg shrimp samples Penaeus vannamei, the sensitivity for V. alginolyticus detection was 2 × 10(3) CFU/g (equivalent to 4 CFU per reaction) while PCR was 10 times less sensitive. The LAMP-LFD method for V. alginolyticus correctly identified 21 isolates of V. alginolyticus but did not recognize 23 non-V. alginolyticus Vibrio isolates and 15 non-Vibrio isolates. In summary, this LAMP-LFD method targeted to the rpoX gene is a convenient assay for specific identification of V. alginolyticus with high sensitivity.
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%