Merck
  • Salicylanilide carbamates: Promising antibacterial agents with high in vitro activity against methicillin-resistant Staphylococcus aureus (MRSA).
Salicylanilide carbamates: Promising antibacterial agents with high in vitro activity against methicillin-resistant Staphylococcus aureus (MRSA).
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-06-17)
Iveta Zadrazilova, Sarka Pospisilova, Martina Masarikova, Ales Imramovsky, Juana Monreal Ferriz, Jarmila Vinsova, Alois Cizek, Josef Jampilek
ABSTRACT
A series of twenty-one salicylanilide N-alkylcarbamates was assessed for novel antibacterial characteristics against three clinical isolates of methicillin-resistant Staphylococcus aureus (MRSA) and S. aureus ATCC 29213 as the reference and quality control strain. The minimum inhibitory concentration was determined by the broth dilution micro-method with subsequent subcultivation of aliquots to assess minimum bactericidal concentration. The bactericidal kinetics was established by time-kill assay. Ampicillin, ciprofloxacin and vancomycin were used as reference antibacterial drugs. All the tested compounds exhibited highly potent anti-MRSA activity (⩽ 0.008-4 μg/mL) comparable or up to 250× higher than that of vancomycin, the standard in the treatment of serious MRSA infections. 4-Chloro-2-(3,4-dichlorophenylcarbamoyl)phenyl butylcarbamate and 4-chloro-2-(3,4-dichlorophenylcarbamoyl)phenyl ethylcarbamate were the most active compounds. In most cases, compounds provided reliable bacteriostatic activity, except for 4-chloro-2-(4-chlorophenylcarbamoyl)phenyl decylcarbamate exhibiting bactericidal effect at 8h (for clinical isolate of MRSA 63718) and at 24h (for clinical isolates of MRSA SA 630 and MRSA SA 3202) at 4× MIC. Structure-activity relationships are discussed.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ampicillin, meets USP testing specifications