Merck
  • Anti-thrombotic effect of rutin isolated from Dendropanax morbifera Leveille.
Anti-thrombotic effect of rutin isolated from Dendropanax morbifera Leveille.
Journal of bioscience and bioengineering (2015-03-18)
Jun-Hui Choi, Dae-Won Kim, Se-Eun Park, Hyo-Jeong Lee, Ki-Man Kim, Kyung-Je Kim, Myung-Kon Kim, Sung-Jun Kim, Seung Kim
ABSTRACT
Dendropanax morbifera H. Lev. is well known in Korean traditional medicine for improvement of blood circulation. In this study, rutin, a bioflavonoid having anti-thrombotic and anticoagulant activities was isolated from a traditional medicinal plant, D. morbifera H. Lev. The chemical characteristics of rutin was studied to be quercetin 3-O-α-l-rhamnopyranosyl-(1-6)-β-d-glucopyranoside using high performance liquid chromatography mass spectrometry (HPLC-MS), proton nuclear magnetic resonance ((1)H NMR) and carbon-13 nuclear magnetic resonance ((13)C NMR). Turbidity and fibrin clotting studies revealed that rutin reduces fibrin clot in concentration dependent manner. Rutin was found to prolong activated partial thromboplastin time (aPTT), prothrombin time (PT) and closure time (CT). Furthermore, it decreased the activity of pro-coagulant protein, thrombin. In vivo study showed that rutin exerted a significant protective effect against collagen and epinephrine (or thrombin) induced acute thromboembolism in mice. These results suggest that rutin has a potent to be an anti-thrombotic agent for cardiovascular diseases.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma