Merck
  • LC-MS/MS analysis of plasma polyunsaturated fatty acids in patients with homozygous sickle cell disease.
LC-MS/MS analysis of plasma polyunsaturated fatty acids in patients with homozygous sickle cell disease.
Clinical and experimental medicine (2014-06-14)
Mutay Aslan, Gamze Celmeli, Filiz Özcan, Alphan Kupesiz
ABSTRACT
The aim of this study was to determine circulating omega-6, omega-3 polyunsaturated fatty acids and prostaglandin E2 (PGE2) levels in steady state sickle cell disease (SCD) patients. Blood was collected from healthy hemoglobin volunteers and steady state homozygous HbSS patients who had not received blood transfusions in the last 3 months. Plasma levels of arachidonic acid (AA, C20:4n-6), dihomo-gamma-linolenic acid (DGLA, C20:3n-6), eicosapentaenoic acid (EPA, C20:5n-3) and docosahexaenoic acid (DHA, C22:6n-3) were determined by an optimized multiple reaction monitoring method using ultrafast liquid chromatography coupled with tandem mass spectrometry. PGE2 was measured in serum samples by enzyme immunoassay. Plasma AA and DGLA were significantly increased while EPA and DHA were significantly decreased in SCD plasma compared to control. Serum PGE2 levels, AA/DHA and AA/EPA ratio was significantly higher in SCD patients when compared to control group. The significant increase in PGE2 levels, AA/EPA and AA/DHA ratio confirms the presence of a proinflammatory state in SCD patients.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
(L)-Dehydroascorbic acid
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Dehydro-L-(+)-ascorbic acid dimer, ≥80% (enzymatic)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis