Merck
  • Evaluation of p-phenylenediamine, o-phenylphenol sodium salt, and 2,4-diaminotoluene in the rat comet assay as part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiated international validation study of in vivo rat alkaline comet assay.
Evaluation of p-phenylenediamine, o-phenylphenol sodium salt, and 2,4-diaminotoluene in the rat comet assay as part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiated international validation study of in vivo rat alkaline comet assay.
Mutation research. Genetic toxicology and environmental mutagenesis (2015-07-28)
Marlies De Boeck, Bas-jan van der Leede, Kathleen De Vlieger, Helena Geys, An Vynckier, Jacky Van Gompel
ABSTRACT
As part of the Japanese Center for the Validation of Alternative Methods (JaCVAM)-initiated international validation study of in vivo rat alkaline comet assay (comet assay), p-phenylenediamine dihydrochloride (PPD), o-phenylphenol sodium salt (OPP), and 2,4-diaminotoluene (2,4-DAT), were analyzed in this laboratory as coded test chemicals. Male Sprague-Dawley rats (7-9 weeks of age) were given three oral doses of the test compounds, 24 and 21 h apart and liver and stomach were sampled 3h after the final dose administration. Under the conditions of the test, no increases in DNA damage were observed in liver and stomach with PPD and OPP up to 100 and 1000 mg/kg/day, respectively. 2,4-DAT, a known genotoxic carcinogen, induced a weak but reproducible, dose-related and statistically significant increase in DNA damage in liver cells while no increases were observed in stomach cells.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)