Merck
  • Probucol Protects Against Asymmetric Dimethylarginine-Induced Apoptosis in the Cultured Human Brain Microvascular Endothelial Cells.
Probucol Protects Against Asymmetric Dimethylarginine-Induced Apoptosis in the Cultured Human Brain Microvascular Endothelial Cells.
Journal of molecular neuroscience : MN (2015-08-10)
Jiwei Ma, Shupeng Zhao, Guojun Gao, Haigang Chang, Pengju Ma, Baozhe Jin
ABSTRACT
Asymmetric dimethylarginine (ADMA) is emerging as a key contributor to endothelial dysfunction. The drug probucol was reported to have an anti-lipid peroxidation effect and improve endothelial dilation function. But little is known about the protective effect of probucol on ADMA-induced human brain microvascular endothelial cell (HBMEC) injury and its underlying mechanisms. Therefore, in this study, we investigated the effect of probucol on ADMA-induced HBMEC injury and its potential mechanisms. Results showed that probucol protected against ADMA-induced HBMEC injury in a dose-dependent manner; probucol pretreatment also significantly reduced the level of reactive oxygen species (ROS) and malondialdehyde (MDA), downregulated the expression of pro-apoptotic gene Bax and caspase-3 activity, as well as increased the brain-derived neurotrophic factor (BDNF) release and promoted anti-apoptotic gene Bcl-2 and eNOS expression in the cultured HBMECs. Furthermore, we found that ADMA significantly increased the phosphorylation of c-Jun NH2-terminal kinase (JNK) and p38, while probucol pretreatment effectively inhibited ADMA-induced JNK and p38 phosphorylation in HBMECs. In conclusion, our present results demonstrated that probucol protected against ADMA-induced HBMEC injury and suppressed oxidative stress through the JNK/p38 MAPK pathway, which was the potential underlying mechanism of HBMEC injury in ischemic cerebrovascular disease.
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
NG,NG-Dimethylarginine dihydrochloride
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%