Merck
Biosynthesis of nectrisine in Thelonectria discophora SANK 18292.
Phytochemistry (2015-04-14)
Ryuki Miyauchi, Toshio Takatsu, Tetsuya Suzuki, Yasunori Ono, Yoichiro Shiba
ABSTRACT
Nectrisine, an iminosugar with a heterocyclic nitrogen-containing 5-membered ring, acts as a glycosidase inhibitor. Thelonectria discophora SANK 18292, a fungus, was identified as a nectrisine producer from its microbial library in our screening for nectrisine producing microorganisms. Biosynthesis of nectrisine produced by the fungus was studied using stable isotope tracer techniques. Incorporation of (13)C-labeled d-ribose and d-xylose into nectrisine was confirmed by mass spectrometry and (13)C NMR spectroscopy, which suggested that these were its precursors. Chromatographic separation of the hot water extract from the culture broth afforded not only nectrisine, but also substantial amounts of 4-amino-4-deoxyarabinitol. Incubation of the latter with the crude enzyme of the fungus at room temp. caused an increase in levels of nectrisine together with a decrease in amounts of the administered potential precursor suggesting that it is a biosynthetic intermediate. From these results, a biosynthetic pathway to nectrisine is proposed via d-xylulose 5-phosphate and 4-amino-4-deoxyarabinitol by the pentose phosphate pathway.
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
D-(−)-Ribose, ≥98%
Sigma-Aldrich
D-(−)-Ribose, 98%
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, suitable for fluorescence, BioReagent, ≥98.0% (HPLC)
Sigma-Aldrich
D-(−)-Ribose, ≥99% (GC)
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, ≥98% (elemental analysis)
Sigma-Aldrich
D-(−)-Ribose, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma