Merck
  • Development and Validation of Two Screening Assays for the Hepatitis C Virus NS3 Q80K Polymorphism Associated with Reduced Response to Combination Treatment Regimens Containing Simeprevir.
Development and Validation of Two Screening Assays for the Hepatitis C Virus NS3 Q80K Polymorphism Associated with Reduced Response to Combination Treatment Regimens Containing Simeprevir.
Journal of clinical microbiology (2015-07-03)
C K S Chui, W W Y Dong, J B Joy, A F Y Poon, W Y Dong, T Mo, C K Woods, C Beatty, H Hew, P R Harrigan, C J Brumme
ABSTRACT
Persons with hepatitis C virus (HCV) genotype 1a (GT1a) infections harboring a baseline Q80K polymorphism in nonstructural protein 3 (NS3) have a reduced virologic response to simeprevir in combination with pegylated interferon-alfa and ribavirin. We aimed to develop, validate, and freely disseminate an NS3 clinical sequencing assay to detect the Q80K polymorphism and potentially other HCV NS3 drug resistance mutations. HCV RNA was extracted from frozen plasma using a NucliSENS easyMAG automated nucleic acid extractor, amplified by nested reverse transcription-PCR, and sequenced using Sanger and/or next-generation (MiSeq) methods. Sanger chromatograms were analyzed using in-house software (RECall), and nucleotide mixtures were called automatically. MiSeq reads were iteratively mapped to the H77 reference genome, and consensus NS3 sequences were generated with nucleotides present at >20% called as mixtures. The accuracy, precision, and sensitivity for detecting the Q80K polymorphism were assessed in 70 samples previously sequenced by an external laboratory. A comparison of the sequences generated by the Sanger and MiSeq methods with those determined by an external lab revealed >98.5% nucleotide sequence concordance and zero discordant calls of the Q80K polymorphism. The results were both highly repeatable and reproducible (>99.7% nucleotide concordance and 100% Q80K concordance). The limits of detection (>2 and ∼5 log10 IU/ml for the Sanger and MiSeq assays, respectively) are sufficiently low to allow genotyping in nearly all chronically infected treatment-naive persons. No systematic bias in the under- or overamplification of minority variants was observed. Coinfection with other viruses (e.g., HIV and hepatitis B virus [HBV]) did not affect the assay results. The two independent HCV NS3 sequencing assays with the automated analysis procedures described here are useful tools to screen for the Q80K polymorphism and other HCV protease inhibitor drug resistance mutations.
Sigma-Aldrich
Cresol red, indicator grade, Dye content 95 %
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M