Merck
  • The Bax inhibitor MrBI-1 regulates heat tolerance, apoptotic-like cell death, and virulence in Metarhizium robertsii.
The Bax inhibitor MrBI-1 regulates heat tolerance, apoptotic-like cell death, and virulence in Metarhizium robertsii.
Scientific reports (2015-05-30)
Yixiong Chen, Zhibing Duan, Peilin Chen, Yanfang Shang, Chengshu Wang
ABSTRACT
Bax inhibitor 1 (BI-1) is a highly conserved protein originally identified as a suppressor of the proapoptotic protein Bax to inhibit cell death in animals and plants. The orthologs of BI-1 are widely distributed in filamentous fungi but their functions remain largely unknown. Herein, we report the identification and characterizations of MrBI-1, an ortholog of BI-1, in the entomopathogenic fungus Metarhizium robertsii. First, we found that MrBI-1 could partially rescue mammalian Bax-induced cell death in yeast. Deletion of MrBI-1 impaired fungal development, virulence and heat tolerance in M. robertsii. We also demonstrated that inactivation of MrBI-1 reduced fungal resistance to farnesol but not to hydrogen peroxide, suggesting that MrBI-1 contributes to antiapoptotic-like cell death via the endoplasmic reticulum stress-signaling pathway rather than the classical mitochondrium-dependent pathway. In particular, we found that unlike the observations in yeasts and plants, expression of mammalian Bax did not lead to a lethal effect in M. robertsii; however, it did aggravate the fungal apoptotic effect of farnesol. The results of this study advance our understanding of BI-1-like protein functions in filamentous fungi.
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%