Merck
The influence of ruthenium on vascular tone.
The Journal of pharmacy and pharmacology (2015-04-14)
Bart Pauwels, Charlotte Boydens, Johan Van de Voorde
ABSTRACT
Over the past few years, ruthenium has been under attention for development of organometallic drugs with various therapeutic applications. Because of its favourable characteristics, ruthenium is perfectly suitable for drug design. Ruthenium-containing complexes exert a wide range of biological effects. However, so far, the influence of ruthenium itself on vascular tone has never been studied. The effect of ruthenium was analysed through organ bath studies measuring isometric tension on mice thoracic aorta. After obtaining a stable contraction plateau, cumulative concentration-response curves of the ruthenium-compounds (RuCl3 , Ruthenium Red, [RuCl2 (CO)3 ]2 and RuCl2 (DMSO)4 ) (30-600 μmol/l) were performed. The effect of RuCl3 after contraction with different contractile agents was evaluated. Furthermore, the influence of ruthenium-containing molecules on endogenous (acetylcholine) and exogenous (sodium nitroprusside) NO-mediated relaxations was determined. All studied ruthenium compounds elicit, to some extent, a decrease of the contraction level. Looking further into the underlying mechanism, we found that RuCl3 relaxes aortic rings only when contracted with norepinephrine. This RuCl3 -induced relaxation can be prevented by the antioxidants ascorbic acid and N-acetyl L-cysteine. In addition, ruthenium compounds may diminish acetylcholine- or sodium nitroprusside-induced relaxations. Ruthenium-containing molecules can influence vascular tone induced by norepinephrine due to oxidative inactivation. Moreover, they can undermine NO-mediated responses. This should be considered when developing ruthenium-containing drugs.
Sigma-Aldrich
Acetylcholine chloride, pkg of 150 mg (per vial)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
N-Acetyl-L-cysteine, suitable for cell culture, BioReagent
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC)
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetylcholine chloride, ≥99% (TLC), free-flowing, Redi-Dri
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture