Merck
  • Interference Reduction in Glucose Detection by Redox Potential Tuning: New Glucose Meter Development.
Interference Reduction in Glucose Detection by Redox Potential Tuning: New Glucose Meter Development.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-07-15)
Seong Je Cho, Chul-Ho Cho, Kwang Bok Kim, Min-Hyoung Lee, Jae Hong Kim, Suho Lee, Jaegeol Cho, Suntae Jung, Dong-Min Kim, Yoon-Bo Shim
ABSTRACT
A new glucose meter was developed employing a novel disposable glucose sensor strip comprising a nicotinamide adenine dinucleotide-glucose dehydrogenase (NAD-GDH) and a mixture of Fe compounds as a mediator. An iron complex, 5-(2,5-di(thiophen-2-yl)-1H-pyrrol-1-yl)-1,10-phenanthroline iron(III) chloride (Fe-PhenTPy), was synthesized as a new mediator for the NAD-GDH system. Due to the high oxidation potential of the mediator, the detection potential was tuned to be more closely fitted toward the enzyme reaction potential, less than 400 mV (vs. Ag/AgCl), by mixing with an additional iron mediator. The impedance spectrometry for the enzyme sensor containing the mixed mediators showed an enhanced charge transfer property. In addition, a new cartridge-type glucose meter was manufactured using effective aligned-electrodes, which showed an enhanced response compared with conventional electrode alignment. The proposed glucose sensor resulted in a wide dynamic range in the concentration range of 30 - 500 mg dL(-1) with a reduced interference effect and a good sensitivity of 0.57 μA mM(-1).
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
L-Ascorbic acid, FCC, FG