Merck
  • Bacterial lipopolysaccharide induces rainbow trout myotube atrophy via Akt/FoxO1/Atrogin-1 signaling pathway.
Bacterial lipopolysaccharide induces rainbow trout myotube atrophy via Akt/FoxO1/Atrogin-1 signaling pathway.
Acta biochimica et biophysica Sinica (2015-09-06)
J E Aedo, A E Reyes, R Avendaño-Herrera, A Molina, J A Valdés
ABSTRACT
Lipopolysaccharide (LPS) is considered as a powerful inducer of muscle atrophy in higher vertebrates due to skeletal muscle cell recognition of the endotoxin and a consequent activation of catabolic signaling pathways. In contrast, there is no evidence of LPS directly inducing skeletal muscle atrophy in lower vertebrates, such as fish. For years it has been assumed that fish are resistant to LPS, mainly due to differences in the key features of toll-like receptor (TLR) signaling pathways when compared with mammals. In this study, we report that the stimulation of cultured rainbow trout (Oncorhynchus mykiss) myotubes with LPS (100 ng/ml) resulted in a transient decrease in the pAkt/Akt ratio, a subsequent reduction in the pFoxO1/FoxO1 ratio, and a significant increase in atrogin-1 transcript expression. Preincubation with polymyxin B, an LPS-neutralizing agent, and 740 Y-P, an agonist of p85-PI3K, blocked the effects of LPS. Additionally, LPS treatment induced an increase in protein ubiquitination and a reduction in myotube diameter, both of which are associated with muscular atrophy that is not observed under polymyxin B and 740 Y-P pretreatments. Finally, rainbow trout myotubes expressed the genes tlr1, tlr3, tlr5m, tlr8a1, tlr8a2, tlr9, and tlr22, with significantly increased expressions of tlr5m and tlr9 under LPS stimulation. These results indicate that LPS is an inducer of fish skeletal muscle atrophy and suggest that TLR5M and TLR9 may play important roles in detecting LPS, which supports for the first time the hypothesis that LPS is a direct inducer of skeletal muscle atrophy in teleost species.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder