Merck
  • Activation of Mas Oncogene-Related G Protein-Coupled Receptors Inhibits Neurochemical Alterations in the Spinal Dorsal Horn and Dorsal Root Ganglia Associated with Inflammatory Pain in Rats.
Activation of Mas Oncogene-Related G Protein-Coupled Receptors Inhibits Neurochemical Alterations in the Spinal Dorsal Horn and Dorsal Root Ganglia Associated with Inflammatory Pain in Rats.
The Journal of pharmacology and experimental therapeutics (2015-07-15)
Dongmei Wang, Peizhong Wang, Jianping Jiang, Qingqin Lv, Xueai Zeng, Yanguo Hong
ABSTRACT
Mas oncogene-related G protein-coupled receptor C (MrgC) is unequally expressed in sensory ganglia and has been shown to modulate pathologic pain. This study investigated the mechanism underlying the effect of MrgC receptors on inflammatory pain. Intrathecal administration of the selective MrgC receptor agonist bovine adrenal medulla 8-22 (BAM8-22) (30 nmol) inhibited complete Freund's adjuvant-evoked hyperalgesia. This was associated with the inhibition of protein kinase C-γ and phosphorylated extracellular signal-regulated protein kinase in the spinal cord and/or dorsal root ganglia (DRG). The complete Freund's adjuvant injection in the hindpaw induced an increase in Gq, but not Gi and Gs, protein in the spinal dorsal horn. This increase was inhibited by the intrathecal administration of BAM8-22. The exposure of DRG cultures to bradykinin (10 μM) and prostaglandin E2 (1 μM) increased the expression of calcitonin gene-related peptide (CGRP) and neuronal nitric oxide synthase in small- and medium-sized neurons as well as the levels of CGRP, aspartate, and glutamate in the cultured medium. The bradykinin/prostaglandin E2-induced alterations were absent in the presence of BAM8-22 (10 nM). These results suggest that the activation of MrgC receptors can modulate the increase in the expression of CGRP and neuronal nitric oxide synthase as well as the release of CGRP and excitatory amino acids in DRG associated with inflammatory pain. This modulation results in the inhibition of pain hypersensitivity by suppressing the expression of Gq protein and protein kinase C-γ and extracellular signal-regulated protein kinase signaling pathways in the spinal cord and/or DRG. The present study suggests that MrgC receptors may be a novel target for relieving inflammatory pain.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture