Merck
  • Inhibition of Hyperpolarization-Activated Cation Current in Medium-Sized DRG Neurons Contributed to the Antiallodynic Effect of Methylcobalamin in the Rat of a Chronic Compression of the DRG.
Inhibition of Hyperpolarization-Activated Cation Current in Medium-Sized DRG Neurons Contributed to the Antiallodynic Effect of Methylcobalamin in the Rat of a Chronic Compression of the DRG.
Neural plasticity (2015-06-24)
Ming Zhang, Wenjuan Han, Jianyong Zheng, Fancheng Meng, Xiying Jiao, Sanjue Hu, Hui Xu
ABSTRACT
Recently several lines of evidence demonstrated that methylcobalamin (MeCbl) might have potential analgesic effect in experimental and clinical studies. However, it was reported that MeCbl had no effect on treating lumbar spinal stenosis induced pain. Thus, the effects of short-term and long-term administration of MeCbl were examined in the chronic compression of dorsal root ganglion (CCD) model. We found that mechanical allodynia was significantly inhibited by a continuous application of high dose and a single treatment of a super high dose of MeCbl. Little is known about mechanisms underlying the analgesia of MeCbl. We examined the effect of MeCbl on the spontaneous activity (SA), the excitability, and hyperpolarization-activated nonselective cation ion current in compressed medium-sized dorsal root ganglion (DRG) neurons using extracellular single fiber recording in vivo and whole-cell patch clamp in vitro. We found that MeCbl significantly inhibited the SA of A-type sensory neurons in a dose-dependent manner and inhibited the excitability of medium-sized DRG neurons. In addition, MeCbl also decreased I h current density in injured medium-sized DRG neurons. Our results proved that MeCbl might exert an analgesic effect through the inhibition I h current and then might inhibit the hyperexcitability of primary sensory neurons under neuropathic pain state.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture