Merck
  • Vanadium(IV) and copper(II) complexes of salicylaldimines and aromatic heterocycles: Cytotoxicity, DNA binding and DNA cleavage properties.
Vanadium(IV) and copper(II) complexes of salicylaldimines and aromatic heterocycles: Cytotoxicity, DNA binding and DNA cleavage properties.
Journal of inorganic biochemistry (2015-04-11)
Isabel Correia, Somnath Roy, Cristina P Matos, Sladjana Borovic, Nataliya Butenko, Isabel Cavaco, Fernanda Marques, Julia Lorenzo, Alejandra Rodríguez, Virtudes Moreno, João Costa Pessoa
ABSTRACT
Five copper(II) complexes, [Cu(sal-Gly)(bipy)](1), [Cu(sal-Gly)(phen)] (2), [Cu(sal-l-Ala)(phen)] (3), [Cu(sal-D-Ala)(phen)] (4), [Cu(sal-l-Phe)(phen)] (5) and five oxidovanadium(IV) complexes, [V(IV)O(sal-Gly)(bipy)] (6), [V(IV)O(sal-Gly)(phen)] (7), [V(IV)O(sal-l-Phe)(H2O)] (8), [V(IV)O(sal-l-Phe)(bipy)] (9), [V(IV)O(sal-l-Phe)(phen)] (10) (sal=salicylaldehyde, bipy=2,2'-bipyridine, phen=1,10-phenanthroline) were synthesized and characterized, and their interaction with DNA was evaluated by different techniques: gel electrophoresis, fluorescence, UV-visible and circular dichroism spectroscopy. The complexes interact with calf-thymus DNA and efficiently cleave plasmid DNA in the absence (only 2 and 5) and/or presence of additives. The cleavage ability is concentration-dependent as well as metal and ligand-dependent. Moreover, DNA binding experiments show that the phen-containing Cu(II) and V(IV)O compounds display stronger DNA interaction ability than the corresponding bipy analogues. The complexes present cytotoxic activity against human ovarian (A2780) and breast (MCF7) carcinoma cells. Cell-growth inhibition (IC50) of compounds 1, 2 and 5 in human promyelocytic leukemia (HL60) and human cervical cancer (HeLa) cells were also determined. The copper complexes show much higher cytotoxic activity than the corresponding vanadium complexes and the reference drug cisplatin (except for the sal-Gly complexes); namely, the phenanthroline copper complexes 2-5 are ca. 10-fold more cytotoxic than cisplatin and more cytotoxic than their bipyridine analogues.
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)