Merck
  • Post-transcriptional regulation of SHANK3 expression by microRNAs related to multiple neuropsychiatric disorders.
Post-transcriptional regulation of SHANK3 expression by microRNAs related to multiple neuropsychiatric disorders.
Molecular brain (2015-11-18)
Su-Yeon Choi, Kaifang Pang, Joo Yeon Kim, Jae Ryun Ryu, Hyojin Kang, Zhandong Liu, Won-Ki Kim, Woong Sun, Hyun Kim, Kihoon Han
ABSTRACT
Proper neuronal function requires tight control of gene dosage, and failure of this process underlies the pathogenesis of multiple neuropsychiatric disorders. The SHANK3 gene encoding core scaffolding proteins at glutamatergic postsynapse is a typical dosage-sensitive gene, both deletions and duplications of which are associated with Phelan-McDermid syndrome, autism spectrum disorders, bipolar disorder, intellectual disability, or schizophrenia. However, the regulatory mechanism of SHANK3 expression in neurons itself is poorly understood. Here we show post-transcriptional regulation of SHANK3 expression by three microRNAs (miRNAs), miR-7, miR-34a, and miR-504. Notably, the expression profiles of these miRNAs were previously shown to be altered in some neuropsychiatric disorders which are also associated with SHANK3 dosage changes. These miRNAs regulated the expression of SHANK3 and other genes encoding actin-related proteins that interact with Shank3, through direct binding sites in the 3' untranslated region (UTR). Moreover, overexpression or inhibition of miR-7 and miR-504 affected the dendritic spines of the cultured hippocampal neurons in a Shank3-dependent manner. We further characterized miR-504 as it showed the most significant effect on both SHANK3 expression and dendritic spines among the three miRNAs. Lentivirus-mediated overexpression of miR-504, which mimics its reported expression change in postmortem brain tissues of bipolar disorder, decreased endogenous Shank3 protein in cultured hippocampal neurons. We also revealed that miR-504 is expressed in the cortical and hippocampal regions of human and mouse brains. Our study provides new insight into the miRNA-mediated regulation of SHANK3 expression, and its potential implication in multiple neuropsychiatric disorders associated with altered SHANK3 and miRNA expression profiles.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Sodium chloride solution, 0.85%
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri