Merck
  • Preparation, anticholinesterase activity, and docking study of new 2-butenediamide and oxalamide derivatives.
Preparation, anticholinesterase activity, and docking study of new 2-butenediamide and oxalamide derivatives.
Journal of enzyme inhibition and medicinal chemistry (2014-11-29)
Kadir Ozden Yerdelen, Mehmet Koca, Zeynep Kasap, Baris Anil
ABSTRACT
Several new oxalamide and 2-butenediamide derivatives have been designed, synthesized and evaluated as the acetyl- and butyryl-cholinesterase inhibitors for Alzheimer's disease. The enzyme inhibitory activity of the synthesized compounds was measured using Ellman's colorimetric method. It was revealed that compound 1a (N,N'-bis-(4-chloro-benzyl)-N,N'-diphenyl-oxalamide) showed maximum activity against BuChE with a half maximal inhibitory concentration (IC50) = 1.86 µM and compound 2a (but-2-enedioic acid bis-[(4-chloro-benzyl)-phenyl-amide]) exhibited optimum AChE (IC50 = 1.51 µM) inhibition with a high-selectivity index. To better understand the enzyme-inhibitor interaction of the most active compounds towards cholinesterase, molecular modelling studies were carried out. Docking simulations revealed that inhibitors 1a and 2a targeted both the catalytic active site and the peripheral anionic site of 1ACJ and 1P0I.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Butyrylthiocholine iodide, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%