Merck
  • Rett syndrome like phenotypes in the R255X Mecp2 mutant mouse are rescued by MECP2 transgene.
Rett syndrome like phenotypes in the R255X Mecp2 mutant mouse are rescued by MECP2 transgene.
Human molecular genetics (2015-01-31)
Meagan R Pitcher, José A Herrera, Shelly A Buffington, Mikhail Y Kochukov, Jonathan K Merritt, Amanda R Fisher, N Carolyn Schanen, Mauro Costa-Mattioli, Jeffrey L Neul
ABSTRACT
Rett syndrome (RTT) is a severe neurodevelopmental disorder that is usually caused by mutations in Methyl-CpG-binding Protein 2 (MECP2). Four of the eight common disease causing mutations in MECP2 are nonsense mutations and are responsible for over 35% of all cases of RTT. A strategy to overcome disease-causing nonsense mutations is treatment with nonsense mutation suppressing drugs that allow expression of full-length proteins from mutated genes with premature in-frame stop codons. To determine if this strategy is useful in RTT, we characterized a new mouse model containing a knock-in nonsense mutation (p.R255X) in the Mecp2 locus (Mecp2(R255X)). To determine whether the truncated gene product acts as a dominant negative allele and if RTT-like phenotypes could be rescued by expression of wild-type protein, we genetically introduced an extra copy of MECP2 via an MECP2 transgene. The addition of MECP2 transgene to Mecp2(R255X) mice abolished the phenotypic abnormalities and resulted in near complete rescue. Expression of MECP2 transgene Mecp2(R255X) allele also rescued mTORC1 signaling abnormalities discovered in mice with loss of function and overexpression of Mecp2. Finally, we treated Mecp2(R255X) embryonic fibroblasts with the nonsense mutation suppressing drug gentamicin and we were able to induce expression of full-length MeCP2 from the mutant p.R255X allele. These data provide proof of concept that the p.R255X mutation of MECP2 is amenable to the nonsense suppression therapeutic strategy and provide guidelines for the extent of rescue that can be expected by re-expressing MeCP2 protein.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O