- Selective Vaporization of Superheated Nanodroplets for Rapid, Sensitive, Acoustic Biosensing.
Selective Vaporization of Superheated Nanodroplets for Rapid, Sensitive, Acoustic Biosensing.
Advanced healthcare materials (2015-06-19)
Rajarshi Chattaraj, Praveena Mohan, Jeremy D Besmer, Andrew P Goodwin
PMID26084414
ABSTRACT
Superheated perfluorocarbon nano-droplets exhibit promise as sensitive acoustic biosensors. Aggregation of biotin-decorated lipid-shelled droplets by streptavidin greatly increases the yield of bubbles formed by ultrasound-induced vaporization. Streptavidin is sensed down to 1 × 10(-13) m, with differentiable signal appearing in as little as two minutes, using a scalable assay without washing, processing, or development steps.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)