Merck
  • A novel micronucleus in vitro assay utilizing human hematopoietic stem cells.
A novel micronucleus in vitro assay utilizing human hematopoietic stem cells.
Toxicology in vitro : an international journal published in association with BIBRA (2015-07-25)
N Kotova, N Hebert, E-L Härnwall, D Vare, C Mazurier, L Douay, D Jenssen, J Grawé
ABSTRACT
The induction of micronucleated reticulocytes in the bone marrow is a sensitive indicator of chromosomal damage. Therefore, the micronucleus assay in rodents is widely used in genotoxicity and carcinogenicity testing. A test system based on cultured human primary cells could potentially provide better prediction compared to animal tests, increasing patient safety while also implementing the 3Rs principle, i.e. replace, reduce and refine. Hereby, we describe the development of an in vitro micronucleus assay based on animal-free ex vivo culture of human red blood cells from hematopoietic stem cells. To validate the method, five clastogens with direct action, three clastogens requiring metabolic activation, four aneugenic and three non-genotoxic compounds have been tested. Also, different metabolic systems have been applied. Flow cytometry was used for detection and enumeration of micronuclei. Altogether, the results were in agreement with the published data and indicated that a sensitive and cost effective in vitro assay to assess genotoxicity with a potential to high-throughput screening has been developed.
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%