Merck
  • Antibiotic delivery by liposomes from prokaryotic microorganisms: Similia cum similis works better.
Antibiotic delivery by liposomes from prokaryotic microorganisms: Similia cum similis works better.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-06-29)
Ilaria Colzi, Anna N Troyan, Brunella Perito, Enrico Casalone, Riccardo Romoli, Giuseppe Pieraccini, Nataša Škalko-Basnet, Alessandra Adessi, Federico Rossi, Cristina Gonnelli, Sandra Ristori
ABSTRACT
To date the effectiveness of antibiotics is undermined by microbial resistance, threatening public health worldwide. Enhancing the efficacy of the current antibiotic arsenal is an alternative strategy. The administration of antimicrobials encapsulated in nanocarriers, such as liposomes, is considered a viable option, though with some drawbacks related to limited affinity between conventional liposomes and bacterial membranes. Here we propose a novel "top-down" procedure to prepare unconventional liposomes from the membranes of prokaryotes (PD-liposomes). These vectors, being obtained from bacteria with limited growth requirements, also represent low-cost systems for scalable biotechnology production. In depth physico-chemical characterization, carried out with dynamic light scattering (DLS) and Small Angle X-ray Scattering (SAXS), indicated that PD-liposomes can be suitable for the employment as antibiotic vectors. Specifically, DLS showed that the mean diameter of loaded liposomes was ∼200-300nm, while SAXS showed that the structure was similar to conventional liposomes, thus allowing a direct comparison with more standard liposomal formulations. Compared to free penicillin G, PD-liposomes loaded with penicillin G showed minimal inhibitory concentrations against E. coli that were up to 16-times lower. Noteworthy, the extent of the bacterial growth inhibition was found to depend on the microorganisms from which liposomes were derived.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Perfluoroheptanoic acid, ≥97.0%