Merck
  • Two-dimensional polyacrylamide gel electrophoresis of Bali bull (Bos javanicus) seminal plasma proteins and their relationship with semen quality.
Two-dimensional polyacrylamide gel electrophoresis of Bali bull (Bos javanicus) seminal plasma proteins and their relationship with semen quality.
Theriogenology (2015-06-30)
Kazhal Sarsaifi, Abd Wahid Haron, Jaya Vejayan, Rosnina Yusoff, Homayoun Hani, Mohamed Ariff Omar, Lai Wei Hong, Nurhusien Yimer, Tan Ying Ju, Abas-Mazni Othman
ABSTRACT
The present study evaluated the relationship between Bali bull (Bos javanicus) seminal plasma proteins and different semen quality parameters. Semen samples from 10 mature Bali bulls were evaluated for conventional semen parameters (general motility, viability, and normal morphology), sperm functionality (acrosome reaction, sperm penetration rate, sperm penetration index), sperm kinetics (computer-assisted semen analysis parameters such as sperm velocity), and sperm morphology (acrosome and membrane integrity). Frozen-thawed semen with higher sperm motility, viability, acrosome integrity, and membrane integrity (P < 0.05) are consistently higher in acrosome reaction and sperm penetration assay. Three bulls showed the highest, four bulls displayed the medium, and the remaining three bulls showed the lowest for all sperm parameters and SPA. The proteome maps of seminal plasma from high-quality and low-quality Bali bulls were also established. Seminal plasma of both high-quality and low-quality Bali bulls was subjected to two-dimensional SDS-PAGE with isoelectric point ranged from 3 to 10 and molecular weight from 10 to 250 kDa. Approximately 116 spots were detected with Blue Silver stain, and of these spots, 29 were selected and identified by MALDI-TOF/TOF-MS/MS. A majority of the proteins visualized in the seminal plasma two-dimensional maps was successfully identified. An essential group of the identified spots represented binder of sperm 1 (BSP1), clusterin, spermadhesin, tissue inhibitor of metalloproteinases 2 (TIMP-2), and phospholipase A2 (PLA2). Other proteins found in high abundance included seminal ribonuclease, serum albumin, cationic trypsin, and peptide similar to β2 microglobulin. Thus, a reference map of Bali bull seminal plasma proteins has been generated for the very first time and can be used to relate protein pattern changes to physiopathologic events that may influence Bali bull reproductive performance.
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
SAFC
Glycine
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Tromethamine, meets USP testing specifications