Merck
  • Psora-4, a Kv1.3 Blocker, Enhances Differentiation and Maturation in Neural Progenitor Cells.
Psora-4, a Kv1.3 Blocker, Enhances Differentiation and Maturation in Neural Progenitor Cells.
CNS neuroscience & therapeutics (2015-05-16)
Yu-Ye Zhou, Guo-Qiang Hou, Song-Wei He, Zhuo Xiao, Hui-Juan Xu, Ya-Tao Qiu, Sheng Jiang, Hui Zheng, Zhi-Yuan Li
ABSTRACT
The self-repair ability of neural progenitor cells (NPCs) has been found to be activated and protected in several therapies helpful in multiple sclerosis (MS), an inflammatory demyelinating disease of the CNS. As a potential therapeutic target in MS, the role of the ion channel Kv1.3 in NPC self-repair has received limited attention. The aim of this study was to explore the effects of a selective Kv1.3 blocker on NPC neuronal differentiation and maturation. A small-molecule selective blocker for Kv1.3, Psora-4, was added to the differentiation medium of cultured mouse NPCs to assess its effect on NPC differentiation efficiency. Both a polypeptide Kv1.3 blocker and Kv1.3-specific RNA interference were used in parallel experiments. Further, the maturity of newborn neurons in the presence of Psora-4 was measured both by morphological analysis and by whole-cell patch clamping. Psora-4 induced a significant increase in the percentage of neurons. Knockdown of Kv1.3 in NPCs also promoted neuronal differentiation. Both morphological and electrophysiological analyses suggested that NPC-derived neurons in the presence of Psora-4 were more mature. Our studies reveal a crucial role for the ion channel Kv1.3 in the regulation of NPC differentiation and maturation, making Psora-4 a promising candidate molecule for MS treatment.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture