Merck
Cold stress-induced ferroptosis involves the ASK1-p38 pathway.
EMBO reports (2017-09-10)
Kazuki Hattori, Hiroyuki Ishikawa, Chihiro Sakauchi, Saki Takayanagi, Isao Naguro, Hidenori Ichijo
ABSTRACT
A wide variety of cell death mechanisms, such as ferroptosis, have been proposed in mammalian cells, and the classification of cell death attracts global attention because each type of cell death has the potential to play causative roles in specific diseases. However, the precise molecular mechanisms leading to cell death are poorly understood, particularly in ferroptosis. Here, we show that continuous severe cold stress induces ferroptosis and the ASK1-p38 MAPK pathway in multiple cell lines. The activation of the ASK1-p38 pathway is mediated by critical determinants of ferroptosis: MEK activity, iron ions, and lipid peroxide. The chemical compound erastin, a potent ferroptosis inducer, also activates the ASK1-p38 axis downstream of lipid peroxide accumulation and leads to ASK1-dependent cell death in a cell type-specific manner. These lines of evidence provide mechanistic insight into ferroptosis, a type of regulated necrosis.
Product Description
Millipore
SB 202190, SB 202190, CAS 152121-30-7, is a potent, reversible, competitive inhibitor of p38. Inhibits p38 phosphorylation of myelin basic protein. Blocks the activity of p38β (Ki = 16 nM; IC50 = 350 nM).
Sigma-Aldrich
Staurosporine, Streptomyces sp., InSolution, ≥97%, 1 mM solution, protein kinase inhibitor
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
Z-VAD-FMK, solid
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, L-glutamine, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Erastin, ≥98% (HPLC)
Sigma-Aldrich
Cantharidin
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
Sigma-Aldrich
Erastin, A cell-permeable piperazinyl-quinazolinone compound that exhibits oncogene-selective lethality.
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells).