Skip to Content
MilliporeSigma

ADVH9AC94DE4

AChemBlock

2-Chloro-4H-thieno[3,2-b]pyrrole-5-carboxylic acid

Synonym(s):

2-chloro-4H-thieno[3,2-b]pyrrole-5-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H4O2N1Cl1S1
CAS Number:
Molecular Weight:
201.63
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H4ClNO2S/c8-6-2-3-5(12-6)1-4(9-3)7(10)11/h1-2,9H,(H,10,11)

Inchi Key

OWJRYNILRVVVFK-UHFFFAOYSA-N

Purity

95.00%

Storage temp.

Store at Room Tem.

Country of Origin

CN



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library