Skip to Content
Merck

BDPH9BCD36BB

Bide Pharmatech

Bismuth(III) trifluoromethanesulfonate

Synonym(s):

bismuth(3+) tris(trifluoromethanesulfonate)

Select a Size

Change View

About This Item

Linear Formula:
C3O9S3F9Bi1
CAS Number:
Molecular Weight:
656.19
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/3CHF3O3S.Bi/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

Inchi Key

NYENCOMLZDQKNH-UHFFFAOYSA-K

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to off-white powder or crystals



Still not finding the right product?


hstatements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library