Skip to Content

BDPH9BCDFD0E

Bide Pharmatech

5-(4-(Di-p-tolylamino)phenyl)thiophene-2-carbaldehyde

Synonym(s):

5-(4-(di-p-tolylamino)phenyl)thiophene-2-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C25H21O1N1S1
CAS Number:
Molecular Weight:
383.51
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C25H21NOS/c1-18-3-9-21(10-4-18)26(22-11-5-19(2)6-12-22)23-13-7-20(8-14-23)25-16-15-24(17-27)28-25/h3-17H,1-2H3

Inchi Key

MUQFSLMELVMDSR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library