Skip to Content
Merck
Get up to 22% off for Pi Day through March 27.Save Now

ENA447957504

Enamine

4-aminocyclohexane-1-sulfonic acid

Synonym(s):

4-aminocyclohexanesulfonic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ENA447957504 is not currently sold in your country.

About This Item

Linear Formula:
C6H13O3N1S1
CAS Number:
Molecular Weight:
179.24
MDL number:
Product ENA447957504 is not currently sold in your country.

Inchi Code

1S/C6H13NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h5-6H,1-4,7H2,(H,8,9,10)

Inchi Key

RFUPCOHASVQTME-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95

Storage temp.

RT

Country of Origin

UA

Physical Form

powder


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library